Learn More
Stoelting™ Mouse Adaptor
SDP
Catalog No. 10000151 Shop All Stoelting Co Products
Change view
Click to view available options
Animal:
Mouse
Catalog No. Animal
10-000-151 Mouse
1 options
Catalog No. 10-000-151 Supplier Stoelting™ Supplier No. 51624
Add to Cart
Add to Cart
Stoelting Mouse Adaptor holds the mouse's head by means of a palate bar and nose clamp, without ear bars. Head angle may be adjusted to achieve a flat skull or other approach angles.
Specifications
For Use With (Application) Neuroscience
Animal Mouse