Learn More
Sino Biological Anti-GPR64 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 107977T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Mouse GPR64, and purified by antigen affinity chromatography.
Catalog No. 50-240-9485