Learn More
Biomatik Corporation Mouse Adiponectin (ADPN) CLIA Kit
Supplier:  Biomatik Corporation EKU08904
Encompass_Preferred
Mouse Adiponectin (ADPN) CLIA Kit for detecting ADPN in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 5.49-4,000pg/ml. Sensitivity: 2.41pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q60994. Target Synonyms: GBP28; ApM1; AdipoQ; Acrp30; ACDC; APM1; C1Q And Collagen Domain Containing; Adipocyte Complement-Related Protein Of 30 KDa; Adipose Most Abundant Gene Transcript 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8597