Learn More
Biomatik Corporation Mouse Adiponectin (ADPN) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU12303
Encompass_Preferred
Mouse Adiponectin (ADPN) Mini sample ELISA Kit for detecting ADPN in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q60994. Target Synonyms: GBP28; ApM1; AdipoQ; Acrp30; ACDC; APM1; C1Q And Collagen Domain Containing; Adipocyte Complement-Related Protein Of 30 KDa; Adipose Most Abundant Gene Transcript 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6482