Learn More
Sino Biological Recombinant Mouse Adiponectin Protein (His Tag)
Supplier:  Sino Biological 50636M08H1
Encompass_Preferred
A DNA sequence encoding the mouse ADIPOQ (NP_033735.3) (Met 1-Asn 247) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-255-3185