Learn More
BIOHIPPO Mouse ADORA2b Microsample
SDP
Catalog No. 5000314890
Encompass
Mouse ADORA2b Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of ADORA2b in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 78 13-5000 pg/mL with a sensitivity of 28 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse ADORA2b Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse ADORA2b Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse ADORA2b biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14890 Supplier BIOHIPPO Supplier No. BHE15210935
Mouse ADORA2b Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of ADORA2b in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 78 13-5000 pg/mL with a sensitivity of 28 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse ADORA2b Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse ADORA2b Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse ADORA2b biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color