Learn More
American Research Products Inc Mouse ADP ribosyl cyclase 1 C
Supplier:  American Research Products Inc CSBEL004929MO
Encompass
Mouse ADP-ribosyl cyclase 1 (CD38) ELISA kit from CUSABIO Detection Range 0 625 ng/mL-40 ng/mL Sensitivity 0 156 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27431