Learn More
Biomatik Corporation Mouse ADP Ribosyltransferase 1 (ART1) ELISA Kit
Supplier:  Biomatik Corporation EKU02164
Encompass_Preferred
Mouse ADP Ribosyltransferase 1 (ART1) ELISA Kit for detecting ART1 in tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.781-50ng/ml. Sensitivity: 0.32ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q60935. Gene ID: 11870. Target Synonyms: CD296; ART2; ARTC1; Mono (ADP-ribosyl) transferase 1; ADP-ribosyltransferase C2 and C3 toxin-like 1; GPI-linked NAD (P) (+)--arginine ADP-ribosyltransferase 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9149