Learn More
American Research Products Inc Mouse AdVanced glycosylation e
Catalog No. 5000328478
Encompass
Mouse AdVanced glycosylation end product-specific receptor (AGER) ELISA kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 81 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-28478 Supplier American Research Products Inc Supplier No. CSBEL001441MOIS
Mouse AdVanced glycosylation end product-specific receptor (AGER) ELISA kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 81 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich