Learn More
Invitrogen™ Mouse aFGF ELISA Kit
Description
Mouse aFGF quantitates mouse aFGF in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant mouse aFGF.
Specifications
Specifications
| Accession Number | P61148 |
| Assay Range | 12-3000 pg/mL |
| Assay Sensitivity | 12 pg/mL |
| Conjugate | HRP |
| Immunoassay Kit Format | Sandwich ELISA Kit |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| Target Species | Mouse |
| For Use With (Equipment) | Absorbance Microplate Reader |
| Gene Alias | acidic eye-derived fibroblast growth factor; acidic eye-derived growth factor II; Acidic fibroblast growth factor; aFGF; beta-endothelial cell growth factor; Dffrx; ECGF; ECGFA; ECGFB; ECGF-beta; EDGF; EDGF II; Endothelial cell growth factor; Endothelial cell growth factor alpha; Endothelial cell growth factor beta; endothelial cell growth factor beta and alpha chains; endothelial cell growth factor, alpha; endothelial cell growth factor, beta; endothelial growth factor; Fam; FGF; FGF1; Fgf-1; FGF1 protein; Fgfa; FGF-alpha; Fibroblast growth factor; Fibroblast growth factor 1; fibroblast growth factor 1 (acidic); Fibroblast growth factor 1 (heparin binding); fibroblast growth factor, acidic; GLIO703; HBGF1; HBGF-1; heparin-binding growth factor; Heparin-binding growth factor 1; H-FGF-a; M-FGF-a; Prostatropin; unnamed protein productacidic eye-derived fibroblast growth factor; acidic eye-derived growth factor II; Acidic fibroblast growth factor; aFGF; beta-endothelial cell growth factor; Dffrx; ECGF; ECGFA; ECGFB; ECGF-beta; EDGF; EDGF II; Endothelial cell growth factor; Endothelial cell growth factor alpha; Endothelial cell growth factor beta; endothelial cell growth factor beta and alpha chains; endothelial cell growth factor, alpha; endothelial cell growth factor, beta; endothelial growth factor; Fam; FGF; FGF1; Fgf-1; FGF1 protein; Fgfa; FGF-alpha; Fibroblast growth factor; Fibroblast growth factor 1; fibroblast growth factor 1 (acidic); Fibroblast growth factor 1 (heparin binding); fibroblast growth factor, acidic; GLIO703; HBGF1; HBGF-1; heparin-binding growth factor; Heparin-binding growth factor 1; H-FGF-a; M-FGF-a; Prostatropin; unnamed protein product; |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.