Learn More
Sino Biological Mouse AGER PAb 100ul (Sino Biological)
Supplier:  Sino Biological 50489RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse AGER (rM AGER; Catalog#50489-M08H; NP_031451.2; Met 1-Ala 342). AGER specific IgG was purified by mouse AGER affinity chromatography.
Catalog No. 50-237-1225