Learn More
American Research Products Inc Mouse Agouti related protein
Catalog No. 5000327594
Encompass
Mouse Agouti-related protein (AGRP) ELISA kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27594 Supplier American Research Products Inc Supplier No. CSBEL001462MO
Mouse Agouti-related protein (AGRP) ELISA kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich