Learn More
Sino Biological Anti-Angiotensinogen Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50337RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse Angiotensinogen / SerpinA8 / AGT (rM Angiotensinogen / SerpinA8 / AGT; Catalog#50337-M08H; P11859; Met 1-Val 477). Angiotensinogen / SerpinA8 / AGT specific IgG was purified by Mouse Angiotensinogen / SerpinA8 / AGT affinity chromatography.
Catalog No. 50-241-0212