Learn More
ACROBiosystems Anti-Human IgG-coupled Magnetic Beads (recommended for MPCLIA)
Catalog No. 502331319
Encompass_Preferred
The Anti-Human IgG-coupled Magnetic Beads are 2.8 µm superparamagnetic particles covalently coupled to a highly affinity Monoclonal mouse anti-human IgG antibody. The beads can be used to capture the human IgG in Chemiluminescence procedures.The antibody reacts with the Fc portion of human IgG heavy chain but not with the Fab portion of human IgG. No antibody was detected against human IgM or IgA, or against non-immunoglobulin serum proteins. The antibody has been tested by ELISA and/or solid-phase adsorbed to ensure minimal cross-reaction with mouse, cynomolgus and bovine serum proteins, but it is not sure if the product cross-react with immunoglobulins from other species.The Anti-Human IgG-coupled Magnetic Beads is easy to capture the human IgG, and the bounded antibody have no activity lost, this ready to use products could greatly save your protein coupling time and hassle, and help us get the best performance and highly reproducible results.
Catalog No. 50-233-1319 Supplier ACROBiosystems Supplier No. MPCA00410MG
The Anti-Human IgG-coupled Magnetic Beads are 2.8 µm superparamagnetic particles covalently coupled to a highly affinity Monoclonal mouse anti-human IgG antibody. The beads can be used to capture the human IgG in Chemiluminescence procedures.The antibody reacts with the Fc portion of human IgG heavy chain but not with the Fab portion of human IgG. No antibody was detected against human IgM or IgA, or against non-immunoglobulin serum proteins. The antibody has been tested by ELISA and/or solid-phase adsorbed to ensure minimal cross-reaction with mouse, cynomolgus and bovine serum proteins, but it is not sure if the product cross-react with immunoglobulins from other species.The Anti-Human IgG-coupled Magnetic Beads is easy to capture the human IgG, and the bounded antibody have no activity lost, this ready to use products could greatly save your protein coupling time and hassle, and help us get the best performance and highly reproducible results.