Learn More
Sino Biological Recombinant Anti-Fetuin A/FETUA Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 50093R002100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Fetuin-A / AHSG (Catalog#50093-M08H; NP_038493.1; Met 1-Ile 345).
Catalog No. 50-240-8114