Learn More
Sino Biological Mouse AHSG MAb 100ul (Sino Biological)
Supplier:  Sino Biological 50093R022100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Fetuin-A / AHSG (rM Fetuin-A / AHSG; Catalog#50093-M08H; NP_038493.1; Met1-Ile345).
Catalog No. 50-237-3014