Learn More
Sino Biological Anti-Fetuin A/FETUA Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50093RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse Fetuin-A / AHSG (rM Fetuin-A / AHSG; Catalog#50093-M08H; NP_038493.1; Met 1-Ile 345). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-2145