Learn More
Sino Biological Anti-EMAPII/AIMP1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50697T52100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse AIMP1 / EMAP II (Catalog#50697-MNAE; Q62386; Ser145-Lys310). AIMP1 / EMAP II specific IgG was purified by Mouse AIMP1 / EMAP II affinity chromatography.
Catalog No. 50-255-3138