Learn More
American Research Products Inc Mouse Alanine Aminotransferase
Catalog No. 5000328434
Encompass
Mouse Alanine Aminotransferase (ALT/GPT) ELISA Kit from CUSABIO Detection Range 0 156 mU/mL-10 mU/mL Sensitivity 0 039 mU/mL Sample serum plasma cell culture supernates cell lysates Method Sandwich
Catalog No. 50-003-28434 Supplier American Research Products Inc Supplier No. CSBE16539M
Mouse Alanine Aminotransferase (ALT/GPT) ELISA Kit from CUSABIO Detection Range 0 156 mU/mL-10 mU/mL Sensitivity 0 039 mU/mL Sample serum plasma cell culture supernates cell lysates Method Sandwich