Learn More
Abcam Mouse Alanine Aminotransferase (ALT) ELISA Kit, 96TEST
Supplier:  Abcam AB28526396TEST
Encompass_Preferred
Mouse Alanine Aminotransferase (ALT) ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Mouse Alanine Aminotransferase (ALT) in Mouse in Plasma, Cell culture supernatant, Serum samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-221-5505