Learn More
Assaypro MOUSE ALBUMIN ELISA KIT
Supplier:  Assaypro EMA32011
Encompass
The AssayMax Mouse Albumin ELISA kit is designed for detection of mouse albumin in urine and cell culture supernatant samples. This assay employs a quantitative sandwich enzyme immunoassay technique that measures mouse albumin in less than 4 hours.
Catalog No. NC0010653