Learn More
Innovative Research Inc Mouse Albumin Purified 1000mg
Catalog No. 502036442
Encompass_Preferred
Mouse Albumin Purified from Innovative Research has been purified from US origin Mouse Plasma using proprietary chromatographic and cold ethanol fractionation methods. This is a salt free lyophilized powder with a purity of >95%.
Catalog No. 50-203-6442 Supplier Innovative Research Inc Supplier No. IMSALB1000MG
Mouse Albumin Purified from Innovative Research has been purified from US origin Mouse Plasma using proprietary chromatographic and cold ethanol fractionation methods. This is a salt free lyophilized powder with a purity of >95%.