Learn More
Sino Biological Anti-CD166/ALCAM Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50005RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse ALCAM / CD166 extracellular domain (rM ALCAM; Catalog#50005-M08H; NP_033785.1; Met 1-Lys 527). ALCAM specific IgG was purified by mouse ALCAM affinity chromatography.
Catalog No. 50-240-8575