Learn More
Biomatik Corporation Mouse Alcohol Dehydrogenase 1 (ADH1) ELISA Kit
Supplier:  Biomatik Corporation EKU02219
Encompass_Preferred
Mouse Alcohol Dehydrogenase 1 (ADH1) ELISA Kit for detecting ADH1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.133ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P00329. Gene ID: 11522. Target Synonyms: ADH1A; Alcohol Dehydrogenase 1A (Class I) Alpha Polypeptide; Alcohol dehydrogenase subunit alpha. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9145