Learn More
Biomatik Corporation Mouse Alkaline Phosphatase (ALP) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU12357
Encompass_Preferred
Mouse Alkaline Phosphatase (ALP) Mini sample ELISA Kit for detecting ALP in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P24823; P09242. Gene ID: 11650; 11647. Target Synonyms: AKP; ALKP; Basic Phosphatase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7668