Learn More
ACROBiosystems Mouse ALPG (ALPPL2) Recombinant Protein, C-terminal His tag, 100 μg
Catalog No. NC2195297
Encompass_Preferred
Recombinant mouse ALPG (ALPPL2) is supplied as an active, glycosylated protein with a C-terminal His tag, expressed in HEK293 cells and validated by MALS and activity QC. Supplied lyophilized as a 100 μg vial and formulated with trehalose for stability, it is intended for research applications requiring active alkaline phosphatase.
  • Active enzyme with specific activity >1000 pmol/min/μg (4-MUP)
  • Expressed in HEK293 cells
  • C-terminal polyhistidine (His) tag
  • Glycosylated; migrates ~60-66 kDa by SDS-PAGE
  • Dimeric under native conditions (~100-122 kDa by SEC-MALS)
  • Purity >95% (SDS-PAGE) and >90% (SEC-MALS)
  • Low endotoxin <1.0 EU/μg
  • Lyophilized with trehalose; store at -20°C to -70°C
missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'
Catalog No. NC2195297 Supplier ACROBiosystems Supplier No. ALGM52H9100UG
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant mouse ALPG (ALPPL2) is supplied as an active, glycosylated protein with a C-terminal His tag, expressed in HEK293 cells and validated by MALS and activity QC. Supplied lyophilized as a 100 μg vial and formulated with trehalose for stability, it is intended for research applications requiring active alkaline phosphatase.
  • Active enzyme with specific activity >1000 pmol/min/μg (4-MUP)
  • Expressed in HEK293 cells
  • C-terminal polyhistidine (His) tag
  • Glycosylated; migrates ~60-66 kDa by SDS-PAGE
  • Dimeric under native conditions (~100-122 kDa by SEC-MALS)
  • Purity >95% (SDS-PAGE) and >90% (SEC-MALS)
  • Low endotoxin <1.0 EU/μg
  • Lyophilized with trehalose; store at -20°C to -70°C
missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'