Learn More
Abcam Mouse Alpha-1-Microglobulin ELISA Kit, 96 Tests
Catalog No. 502712119
Encompass_Preferred
Mouse Alpha-1-Microglobulin ELISA Kit is a single-wash 90-min Simplestep used to quantify Mouse Alpha-1-Microglobulin with a sensitivity of 1.932 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Design your own immunoassay: we also offer the conjugation-ready antibody pair
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-271-2119 Supplier Abcam Supplier No. AB31625696TESTS
Mouse Alpha-1-Microglobulin ELISA Kit is a single-wash 90-min Simplestep used to quantify Mouse Alpha-1-Microglobulin with a sensitivity of 1.932 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Design your own immunoassay: we also offer the conjugation-ready antibody pair
The product is subject to the following: Abcam Restricted Use Statement