Learn More
Sino Biological Mouse Alpha 2 Antiplasmin/SerpinF2 Matched ELISA Antibody Pair Set 5 Plates
Catalog No. 5000785724
Encompass_Preferred
The Mouse SerpinF2 ELISA Pair Set is for the quantitative determinationof Mouse SerpinF2.This ELISA Pair Set contains the basic components required for the development of sandwich ELISAs.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for SerpinF2 coated on a 96-well plate. Standards and samples are added to the wells, and any SerpinF2 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-SerpinF2 polyclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of SerpinF2 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-85724 Supplier Sino Biological Supplier No. SEK501675
The Mouse SerpinF2 ELISA Pair Set is for the quantitative determinationof Mouse SerpinF2.This ELISA Pair Set contains the basic components required for the development of sandwich ELISAs.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for SerpinF2 coated on a 96-well plate. Standards and samples are added to the wells, and any SerpinF2 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-SerpinF2 polyclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of SerpinF2 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.