Learn More
American Research Products Inc Mouse Alpha fetoprotein AFP E
Catalog No. 5000329415
Encompass
Mouse Alpha-fetoprotein AFP ELISA Kit from CUSABIO Detection Range 1 23 ng/ml-900 ng/ml Sensitivity 1 23 ng/ml Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29415 Supplier American Research Products Inc Supplier No. CSBE08282M
Mouse Alpha-fetoprotein AFP ELISA Kit from CUSABIO Detection Range 1 23 ng/ml-900 ng/ml Sensitivity 1 23 ng/ml Sample serum plasma tissue homogenates Method Sandwich