Learn More
Abcam Mouse alpha Macroglobulin ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB1088871X96TESTS
Encompass_Preferred
Mouse alpha Macroglobulin ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Mouse alpha Macroglobulin in Mouse in Urine, Plasma, Cell culture supernatant, Serum samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2590272