Learn More
Biomatik Corporation Mouse Alpha-Hemoglobin Stabilizing Protein (aHSP) ELISA Kit
Supplier:  Biomatik Corporation EKU10627
Encompass_Preferred
Mouse Alpha-Hemoglobin Stabilizing Protein (aHSP) ELISA Kit for detecting aHSP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 13.8pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9CY02. Gene ID: 170812. Target Synonyms: A-HSP; ERAF; EDRF; Erythroid Associated Factor; Erythroid differentiation-related factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8323