Learn More
Biomatik Corporation Mouse Alpha-Tocopherol Transfer Protein (TTPa) ELISA Kit
Supplier:  Biomatik Corporation EKU02258
Encompass_Preferred
Mouse Alpha-Tocopherol Transfer Protein (TTPa) ELISA Kit for detecting TTPa in tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8BWP5. Gene ID: 50500. Target Synonyms: AVED; TTP1; alphaTTP; Ataxia (Friedreich-Like) With Vitamin E Deficiency. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6825