Learn More
Biomatik Corporation Mouse ALT (Alanine Transaminase) ELISA Kit
Supplier:  Biomatik Corporation EKF58592
Encompass_Preferred
Mouse ALT (Alanine Transaminase) ELISA Kit for detecting ALT in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q8QZR5. Target Synonyms: ALT; Alanine Transaminase; ALAT; SGPT; alanine aminotransferase; serum glutamic pyruvic transaminase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9169