Learn More
American Research Products Inc Mouse Aminopeptidase N ANPEP
Catalog No. 5000326899
Encompass
Mouse Aminopeptidase N (ANPEP) ELISA kit from CUSABIO Detection Range 39 mU/mL-2500 mU/mL Sensitivity 9 7 mU/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26899 Supplier American Research Products Inc Supplier No. CSBEL001827MO
Mouse Aminopeptidase N (ANPEP) ELISA kit from CUSABIO Detection Range 39 mU/mL-2500 mU/mL Sensitivity 9 7 mU/mL Sample serum plasma tissue homogenates Method Sandwich