Learn More
Abcam Mouse and Rabbit Specific HRP/AEC IHC Detection Kit - Micro-polymer, 15ML
Catalog No. 502586408
Encompass_Preferred
This IHC detection kit is used to detect mouse and rabbit primary antibodies in immunohistochemistry (IHC).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-6408 Supplier Abcam Supplier No. AB23646715ML
This IHC detection kit is used to detect mouse and rabbit primary antibodies in immunohistochemistry (IHC).
The product is subject to the following: Abcam Restricted Use Statement