Learn More
Alpco Ltd Mouse / Rat Progesterone ELISA
SDP
Supplier:  Alpco Ltd 55PROMSE01
Encompass_Preferred
The Mouse / Rat Progesterone ELISA is a competitive immunoassay for the quantitative measurement of progesterone in rat and mouse serum or plasma.
  • Includes ready to use standards
  • Requires only 10 μL sample per well
  • Short incubation time of 1.5 hours
  • See catalog number 55-DEV99RC for kit controls
Research Use Only. Not for Use in Diagnostic Procedures.
Catalog No. 50-305-145