Learn More
World Precision Instrument Mouse Gas Anesthesia Mask
SDP
Catalog No. NC2012935
Encompass
WPI's 505226 is compatible for use with the 505313 mouse stereotaxic instruments including:  Mouse and Neonatal Rat Adaptor (Product Number 505243), Just for Mouse Stereotaxic Series (Product Number 505313) and Just for Mice Stereotaxic (Product Number 505243) Series.  
Adjustable snout clamp provides secure positioning for direct gas delivery.  Inlet/outlet and outlet tubing connections allow for continuous flow of anesthesic gas and active gas scavenging from gas scavenger as shown in the image below.  
Catalog No. NC2012935 Supplier World Precision Instrument Supplier No. 505225
WPI's 505226 is compatible for use with the 505313 mouse stereotaxic instruments including:  Mouse and Neonatal Rat Adaptor (Product Number 505243), Just for Mouse Stereotaxic Series (Product Number 505313) and Just for Mice Stereotaxic (Product Number 505243) Series.  
Adjustable snout clamp provides secure positioning for direct gas delivery.  Inlet/outlet and outlet tubing connections allow for continuous flow of anesthesic gas and active gas scavenging from gas scavenger as shown in the image below.  