Learn More
Raybiotech Inc C-Series Mouse Angiogenesis Antibody Array 1 Kit.
Supplier:  Raybiotech Inc AAMANG14
Encompass_Preferred
Detects 24 Mouse Angiogenic Factors. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: Eotaxin-1 (CCL11), Fas Ligand (TNFSF6), bFGF, GCSF, GM-CSF, IFN-gamma, IGF-2, IL-1 alpha (IL-1 F1), IL-1 beta (IL-1 F2), IL-12 p40/p70, IL-12 p70, IL-13, IL-6, IL-9, Leptin, MCP-1 (CCL2), M-CSF, MIG (CXCL9), Platelet Factor 4 (CXCL4), TIMP-1, TIMP-2, TNF alpha, Thrombopoietin (TPO), VEGF-A
Catalog No. 50-194-8670