Learn More
Biomatik Corporation Mouse Angiogenic Factor With G Patch And FHA Domains 1 (AGGF1) ELISA Kit
Supplier:  Biomatik Corporation EKU02349
Encompass_Preferred
Mouse Angiogenic Factor With G Patch And FHA Domains 1 (AGGF1) ELISA Kit for detecting AGGF1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q7TN31. Gene ID: 66549. Target Synonyms: GPATC7; GPATCH7; VG5Q; Angiogenic factor VG5Q; G patch domain-containing protein 7; Vasculogenesis gene on 5q protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5644