Learn More
ACROBiosystems ACROBiosystems Mouse Angiopoietin-like 3 / ANGPTL3 Protein, His Tag
Catalog No. 502106904
Encompass_Preferred
Recombinant Protein;100UG;Mouse Angiopoietin-like 3, His Tag (AN3-M52H3) is expressed from human 293 cells (HEK293). It contains AA Ser 17 - Thr 455 (Accession # Q9R182-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 52.6 kDa.
Catalog No. 50-210-6904 Supplier ACROBiosystems Supplier No. AN3M52H3100UG
Recombinant Protein;100UG;Mouse Angiopoietin-like 3, His Tag (AN3-M52H3) is expressed from human 293 cells (HEK293). It contains AA Ser 17 - Thr 455 (Accession # Q9R182-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 52.6 kDa.