Learn More
American Research Products Inc Mouse Angiopoietin related pro
Supplier:  American Research Products Inc CSBEL001715MO
Encompass
Mouse Angiopoietin-related protein 7 (ANGPTL7) ELISA kit from CUSABIO Detection Range 6 25 pg/mL-400 pg/mL Sensitivity 1 56 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27416