Learn More
American Research Products Inc Mouse Angiopoietin related pro
Supplier: American Research Products Inc CSBEL001714MO
Mouse Angiopoietin-related protein 6 (ANGPTL6) ELISA kit from CUSABIO Detection Range 62 5 pg/mL-4000 pg/mL Sensitivity 15 6 pg/mL Sample serum plasma tissue homogenates Method Sandwich