Learn More
American Research Products Inc Mouse Angiopoietin related pro
Catalog No. 5000327553
Encompass
Mouse Angiopoietin-related protein 2 (ANGPTL2) ELISA kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-27553 Supplier American Research Products Inc Supplier No. CSBEL001710MO
Mouse Angiopoietin-related protein 2 (ANGPTL2) ELISA kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich