Learn More
American Research Products Inc Mouse Angiopoietin related pro
Supplier: American Research Products Inc CSBEL001711MO
Mouse Angiopoietin-related protein 3 (ANGPTL3) ELISA kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 10 24 pg/mL Sample serum plasma tissue homogenates Method Sandwich