Learn More
Biomatik Corporation Mouse Angiopoietin-Related Protein 3 (Angptl3) Elisa Kit
Supplier:  Biomatik Corporation EKC31374
Encompass_Preferred
Mouse Angiopoietin-Related Protein 3 (Angptl3) Elisa Kit for detecting Angiopoietin-Like 3 in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.25-2000pg/ml. Sensitivity: 10.24pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9R182; Angptl3. Target Synonyms: Angptl3Angiopoietin-related protein 3; Angiopoietin-like protein 3) [Cleaved into: ANGPTL3 (17-224)]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5669