Learn More
Biomatik Corporation Mouse Angiotensin I Converting Enzyme (ACE) CLIA Kit
Supplier:  Biomatik Corporation EKU08771
Encompass_Preferred
Mouse Angiotensin I Converting Enzyme (ACE) CLIA Kit for detecting ACE in serum, plasma, tissue homogenates, cerebrospinal fluid, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 54.9-40,000pg/ml. Sensitivity: 21.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P09470. Gene ID: 11421. Target Synonyms: CD143; ACE1; DCP1; ACEI; ACE-I; Kininase II; Angiotensin-Converting Enzyme; Peptidyl-Dipeptidase A; Dipeptidyl Carboxypeptidase 1; Angiotensin-converting enzyme, soluble form. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6087