Learn More
Biomatik Corporation Mouse Angiotensin III (AngIII) ELISA Kit
Supplier:  Biomatik Corporation EKU10636
Encompass_Preferred
Mouse Angiotensin III (AngIII) ELISA Kit for detecting AngIII in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 12.35-1,000pg/ml. Sensitivity: 4.42pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P97802. Gene ID: 11730. Target Synonyms: Ang-III; Angiotensin-3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8320