Learn More
Sino Biological Mouse ANGPT2 Protein 500ug (Sino Biological)
Supplier:  Sino Biological 50298M07H500
Encompass_Preferred
A DNA sequence encoding the mouse ANGPT2 (NP_031452.2) (Lys275-Phe496) was expressed with a N-terminal polyhistidine tag.
Catalog No. 50-237-0699