Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse ANGPTL1/Angiopoietin-3 ELISA Kit

Catalog No. EM7RBX5
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EM7RBX5 5 x 96 Tests
EM7RB 96 Tests
EM7RBX10 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. EM7RBX5 Supplier Invitrogen™ Supplier No. EM7RBX5
Get Up to $3,000 in Extra Life Sciences Consumables with Purchase
View all promotions
Add to Cart
Add to Cart

ELISA

Mouse ANGPTL1/Angiopoietin-3 quantitates mouse ANGPTL1/Angiopoietin-3 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant mouse ANGPTL1/Angiopoietin-3.

Angiopoietins are members of the vascular endothelial growth factor family and the only known growth factors largely specific for vascular endothelium. Angiopoietin-1, angiopoietin-2, and angiopoietin-4 participate in the formation of blood vessels. The protein encoded by this gene is another member of the angiopoietin family that is widely expressed in adult tissues with mRNA levels highest in highly vascularized tissues. This protein was found to be a secretory protein that does not act as an endothelial cell mitogen in vitro.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q640P2
Assay Range 0.85-200 ng/mL
Assay Sensitivity 0.85 ng/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Mouse
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias 2810039D03Rik; ANG3; ANG-3; angioarrestin; angiopoietin 3; angiopoietin like 1; angiopoietin Y1; angiopoietin-3; angiopoietin-like 1; angiopoietin-like protein 1; Angiopoietin-related protein 1; ANGPT3; ANGPTL1; AngY; ARP1; dJ595C2.2; dJ595C2.2 (angiopoietin Y1); PSEC0154; UNQ162; UNQ162/PRO1882810039D03Rik; ANG3; ANG-3; angioarrestin; angiopoietin 3; angiopoietin like 1; angiopoietin Y1; angiopoietin-3; angiopoietin-like 1; angiopoietin-like protein 1; Angiopoietin-related protein 1; ANGPT3; ANGPTL1; AngY; ARP1; dJ595C2.2; dJ595C2.2 (angiopoietin Y1); PSEC0154; UNQ162; UNQ162/PRO188;
Gene ID (Entrez) 72713
Gene Symbol Angptl1
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target ANGPTL1
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.