Learn More
Raybiotech Inc Mouse ANGPTL3 ELISA Kit
Supplier:  Raybiotech Inc ELMANGPTL31
Encompass_Preferred
For cell culture supernatants, EDTA treated plasma, and serum samples. Heparin and citrate are not recommended. Sensitivity: 40 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: Cross Reactivity: This ELISA kit shows no cross-reactivity with the following cytokines tested: Mouse 6Ckine, Activin A, ADAMTS1, Adiponectin, Angiopoietin-3, ANGTL-4 Artemin, CCL28, CD36, Chordin, CRP, E-Cadherin, Epigen, Epiregulin, Fas, Galectin-7, gp130, Granzyme B, Gremlin, IFN-R1, IL-17B, IL-17B R, IL-22, MIP-1b, MMP-2, MMP-3, MMP-10, PDGF-AA, Persephin, sFRP-3, Shh-N, SLAM, TCK-1, TECK, TGFb1, TNF RI, TNF RII, TPO, TRANCE, TremL1, TWEAK, VEGF-B, VEGF-R2, VCAM-1, VEGF.
Catalog No. 50-194-9925